Anti-NFKBIB Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP13780BEA 350 EUR
ABGEAP13780B
Anti-NFKBIB Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NFKBIB (nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor, beta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR