Anti-PRMT5 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP13773AEA 350 EUR
ABGEAP13773A
Anti-PRMT5 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PRMT5 (protein arginine methyltransferase 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR