Anti-SF3B1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP13754AEA 360 EUR
ABGEAP13754A
Anti-SF3B1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SF3B1 (splicing factor 3b, subunit 1, 155kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR