Anti-TNFRSF10D Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP13742BEA 350 EUR
ABGEAP13742B
Anti-TNFRSF10D Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TNFRSF10D (tumor necrosis factor receptor superfamily, member 10d, decoy with truncated death domain)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR