Anti-PRSS2 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP13719CEA 350 EUR
ABGEAP13719C
Anti-PRSS2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PRSS2 (protease, serine, 2 (trypsin 2))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR