Anti-SUMO1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP13711AEA 360 EUR
ABGEAP13711A
Anti-SUMO1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SUMO1 (small ubiquitin-like modifier 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR