Anti-FOLH1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP13707AEA 350 EUR
ABGEAP13707A
Anti-FOLH1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: FOLH1 (folate hydrolase (prostate-specific membrane antigen) 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR