Anti-TNFRSF10A Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP13702BEA 360 EUR
ABGEAP13702B
Anti-TNFRSF10A Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TNFRSF10A (tumor necrosis factor receptor superfamily, member 10a)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR