Anti-GRM1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP13701BEA 350 EUR
ABGEAP13701B
Anti-GRM1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GRM1 (glutamate receptor, metabotropic 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR