Anti-HLA-DRB4 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP13690AEA 348 EUR
ABGEAP13690A
Anti-HLA-DRB4 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: HLA-DRB4 (major histocompatibility complex, class II, DR beta 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR