Anti-ATOH8 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP13675AEA 348 EUR
ABGEAP13675A
Anti-ATOH8 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ATOH8 (atonal homolog 8 (Drosophila))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR