Anti-VKORC1L1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP13669AEA 348 EUR
ABGEAP13669A
Anti-VKORC1L1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: VKORC1L1 (vitamin K epoxide reductase complex, subunit 1-like 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR