Anti-EMILIN3 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP13662BEA 350 EUR
ABGEAP13662B
Anti-EMILIN3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: EMILIN3 (elastin microfibril interfacer 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR