Anti-ANKRD22 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP13661BEA 348 EUR
ABGEAP13661B
Anti-ANKRD22 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ANKRD22 (ankyrin repeat domain 22)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR