Anti-B4GALNT3 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP13658CEA 350 EUR
ABGEAP13658C
Anti-B4GALNT3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: B4GALNT3 (beta-1,4-N-acetyl-galactosaminyl transferase 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR