Cell culture flasks, for cell suspension, BD Falcon™
Lieferant: BD Biosciences
Corning®
734-0962EA 255.78 EUR
734-0962
Cell culture flasks, for cell suspension, BD Falcon™
Kolben Zellkulturkolben
These flasks are specifically designed for automated cell culture. Cell culture Multi-Flasks enable you to grow more cells faster and easier, thereby making your cell culture workflow more productive.
  • With white plug seal screw cap
  • Secure stacking provides stability from accidental tips or spills, and protection against contamination (canted neck flasks)
  • Nonpyrogenic tested to less than 0.1 EU/mL
  • Volumetric graduations and writing patch
  • Growth area and volume are nominal