Bosshead, square
Lieferant: Juchheim
JUCHVL04809VAEA 10.5 EUR
JUCHVL04809VA
Bosshead, square
Klemmen Stativmuffen
Bosshead made from tempered cast iron, epoxy coated, with stainless steel hex socket (M6).