Pipette tips, hinged rack
Lieferant: Corning Life Sciences
Corning®
613-5500EA 101.45 EUR
CORN4712 CORN4714 CORN4715 CORN4710 CORN4711 613-5500
Pipette tips, hinged rack
Pipettenspitzen
These universal tips are designed to assure a reliable fit with all major brand pipettors. They are available in two sizes: 200 µl and 1000 µl.
  • Hinged rack allows easy rack stacking and steam penetration during autoclaving
  • Racks can be reused: Simple replace the rack insert with a new one
  • Certified RNase- and DNase-free; non pyrogenic
  • Replacement inserts reduce waste