Trichter, ohne Stiel
Lieferant: Corning
PYREX® Corning®
CORN6240-100EA 0 EUR
CORN6240-75 CORN6240-100
Trichter, ohne Stiel
Trichter Filtertrichter Filtertrichter
PYREX®, borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
These stemless funnels are of molded construction with bottoms ground flat and have a 6 mm opening. For use in the sugar industries.