Handheld refractometer, Oechsle
Lieferant: Euromex
635-0313EA 156 EUR
635-0313
Handheld refractometer, Oechsle
Refraktometer Handrefraktometer
Digital refractometer designed to measure sugar concentrations in grapes and fruits.
  • Automated temperature compensation (ATC)
  • Calibration at a temperature of 20 °C
  • Requires only a few drops of liquid
Handheld refractometer designed for measurement of sugar concentrations of grape must and fruits.