UV digital photodocumentation system
EDVO555EA 1570 EUR
EDVO555
UV digital photodocumentation system
Both the midrange UV transilluminator (model 558) and the photodocumentation system (model 551) together!
The hood accommodates gels up to 9,5×11 cm. Photos may be downloaded to a computer.
The midrange UV transilluminator is designed to visualise DNA stained with ethidium bromide and other fluorescent stains.