Tubing connectors, T-shaped
Lieferant: Semadeni
229-4632EA 27.7 EUR
229-4632 229-4631 229-4633
Tubing connectors, T-shaped
Verbinder Schlauchverbinder
TPX, transparent.
  • Heat resistance up to +130 °C
  • Sterilisable