Sound level meter
Lieferant: Breukhoven
BREU6662315EA 0 EUR
BREU6662315
Sound level meter
Portable digital sound level meter.
  • Digital display with analogue graph
  • Auto/Manual ranging
  • Max Hold, F/S response, A/C weighting
  • Analog output