Microtubes, 1,5 ml, secure-lock
Lieferant: Semadeni
SEMA1580EA 29.9 EUR
SEMA1580
Microtubes, 1,5 ml, secure-lock
Röhrchen Reaktionsgefäße
PP, translucent.
  • With writing area
  • Very good chemical resistance
  • Very good temperature resistance
With Secure-lock.