Microwave boxes
Lieferant: Semadeni
SEMA3736EA 7.5 EUR
SEMA3736
Microwave boxes
Boxen Lagerboxen
PP, translucent, with red lid.
  • Suitable for freezing down to –40 °C
  • With small, closable vent in the lid for pressure equalisation when heated
  • Excellent chemical resistance, temperature resistance up to +110 °C
  • Approved for food contact
From particularly microwave permeable plastic, machine washable.