Filter cartridges
Lieferant: BartelsRieger Atemschutztechnik
BART924000EA 41.1 EUR
BART924000
Filter cartridges
Filter Filterkartuschen
The combination filter 39 ASt is designed for use in areas contaminated by dusts and solvent vapours.
  • Combined filters combine the protection of a gas and particle filter in one product
  • The design guarantees the filtration of organic gases and vapours (boiling point >65 °C) and a particle separation efficiency >94,95%
Cartridge filters can only be used in combination with a filter adapter on face pieces with thread connector connection.
Standards: According to DIN EN 14387:2008, Class A1 P2 RD. CE certified.