Sterile swab
Lieferant: Hach
HACH2554300EA 46.3 EUR
HACH2554300
Sterile swab
Tupfer Tupfer für die Probenahme
Sterile swab used to transfer colonies from positive membrane filtration plates to confirmation broth.
  • Length:152,4 mm