Microinjection capillaries, Biopsy Tip II
Lieferant: Eppendorf
EPPE5195000.060EA 489 EUR
EPPE5195000.060
Microinjection capillaries, Biopsy Tip II
Mikroinjektionskapillaren
Biopsy Tip II, capillary for laser-assisted biopsy of cells and organelles.
  • Flange length 0,5 mm
  • Rounded tip end to minimise oolemma trauma
  • 20 µm inner diameter
  • 15° tip angle
  • Sterilised by gamma irradiation
  • Non cytotoxic (as proven by mouse embryo assay)
  • Capillary safe for optimum protection