5-Amino-1-methyl-1H-pyrazol

Lieferant: SIA ENAMINE

ENAMEN300-14223.1GEA 0 EUR
ENAMEN300-14223.1G
5-Amino-1-methyl-1H-pyrazol
5-Amino-1-methyl-1H-pyrazole
Formel: C₄H₇N₃
Molecular Weight: 97,12 g/mol
MDL: MFCD00068156
CAS-Nummer: 1192-21-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR