Hafnium(IV)ethanolat ≥99%

Lieferant: ABCR GMBH
Achtung

ABCRAB106071.0005EA 0 EUR
ABCRAB106071.0005
Hafnium(IV)ethanolat ≥99%
Hafnium(IV)ethanolat

Alkoxide, Metal Alkoxides, Volatile Precursors for CVD

Formel: C₈H₂₀HfO₄
Molecular Weight: 358,732 g/mol
MDL: MFCD00070459
CAS-Nummer: 13428-80-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR