2-Aminophenol-4-sulfonamid

Lieferant: SIA ENAMINE

ENAMEN30001211.1GEA 0 EUR
ENAMEN30001211.5G ENAMEN30001211.10G ENAMEN30001211.1G
2-Aminophenol-4-sulfonamid
2-Aminophenol-4-sulfonamid
Formel: C₆H₈N₂O₃S
MDL: MFCD00035782
CAS-Nummer: 98-32-8
EINECS: 202-657-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR