Benzyltrimethylammoniumtribromid

Lieferant: Molekula

MOLE70318276-1GEA 0 EUR
MOLE70318276-1G
Benzyltrimethylammoniumtribromid
Benzyltrimethylammoniumtribromid
Formel: C₁₀H₁₆Br₃N
Molecular Weight: 389,97
MDL: MFCD00134423
CAS-Nummer: 111865-47-5
EINECS: 000-000-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR