(Trimethylstannyl)butyldimethylsilan
Lieferant: SILANE (PART OF CHEMPUR)
|
Gefahr
|
SILNSILS20410.5EA
0
EUR
SILNSILS20410.5
SILNSILS20410.1
(Trimethylstannyl)butyldimethylsilan
(Trimethylstannyl)butyldimethylsilan
Formel:
C₉H₂₄SiSn |
MDL:
MFCD03093828 CAS-Nummer: 103731-29-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...