1-(4-Methoxyphenyl)piperazin Dihydrochlorid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIALM23004-25GEA 112 EUR
SIALM23004-25G
1-(4-Methoxyphenyl)piperazin Dihydrochlorid, Sigma-Aldrich®
1-(4-Methoxyphenyl)piperazin Dihydrochlorid
Formel: C₁₁H₁₈Cl₂N₂O
Molecular Weight: 265,18 g/mol
MDL: MFCD00012767
CAS-Nummer: 38869-47-5
EINECS: 254-166-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR