4-(Isobutylcarbamoyl)phenylboronsäure

Lieferant: COMBI-BLOCKS INC.

COBBBB-3011-1GEA 143 EUR
COBBBB-3011-1G COBBBB-3011-5G
4-(Isobutylcarbamoyl)phenylboronsäure
4-(Isobutylcarbamoyl)phenylboronsäure
Formel: C₁₁H₁₆BNO₃
Molecular Weight: 221,06
MDL: MFCD03411955
CAS-Nummer: 850568-13-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR