5-Amino-2-brombenzoesäure 95%
Lieferant: Thermo Scientific
|
Achtung
|
H60382.06EA
78.7
EUR
H60382.06
H60382.14
5-Amino-2-brombenzoesäure 95%
5-Amino-2-brombenzoesäure
Formel:
C₇H₆BrNO₂ Molecular Weight: 216,03 Schmelzpunkt: 171…179 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07368968 CAS-Nummer: 2840-02-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...