2-Amino-3-chlor-5-(trifluormethyl)pyridin, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ200
SIAL366080-25GEA
39.3
EUR
SIAL366080-25G
2-Amino-3-chlor-5-(trifluormethyl)pyridin, Sigma-Aldrich®
2-Amino-3-chloro-5-(trifluoromethyl)pyridine
Formel:
C₆H₄ClF₃N₂ Molecular Weight: 196,5600 g/mol |
MDL:
MFCD00042154 CAS-Nummer: 79456-26-1 EINECS: 401-670-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...