7-(2,3-Dihydroxypropyl)theophyllin

Lieferant: U.S. Pharmacopeia

USPH1231502EA 332 EUR
USPH1231502
7-(2,3-Dihydroxypropyl)theophyllin
7-(2,3-Dihydroxypropyl)theophyllin
Formel: C₁₀H₁₄N₄O₄
Molecular Weight: 254,24 g/mol
MDL: MFCD00005759
CAS-Nummer: 479-18-5
EINECS: 207-526-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR