trans-1,4-Cyclohexandicarbonsäure 95%
|
Achtung
|
H60089.03EA
36.19
EUR
H60089.03
H60089.06
trans-1,4-Cyclohexandicarbonsäure 95%
trans-1,4-Cyclohexanedicarboxylic acid
Formel:
C₈H₁₂O₄ Molecular Weight: 172,18 g/mol Schmelzpunkt: >300 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00066203 CAS-Nummer: 619-82-9 EINECS: 210-614-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...