Hepatocyte culture medium
Lieferant: Corning Life Sciences
Corning®
734-2386EA 243.77 EUR
734-2386
Hepatocyte culture medium
Zellkulturmedien
Hepatocyte culture medium is a serum-free, fully-defined medium optimised for hepatocyte differentiation.
Lieferumfang: Kit comprises hepatocyte culture medium (500 ml) and EGF (5 µg).