Endothelial culture medium
Lieferant: Corning Life Sciences
Corning®
734-2385EA 421.65 EUR
734-2385
Endothelial culture medium
Zellkulturmedien
Endothelial culture medium is optimised to promote rapid growth (3- to 5-fold increase in cell number over five days) of endothelial cells from a variety of sources.
Lieferumfang: Kit comprises endothelial cell culture medium (500 ml), epidermal growth factor (5 µg), endothelial cell growth supplement (100 mg), and trypsin inhibitor (50 mg).