Flammable atmospheres detection device, Dräger X-am 2500 EX
Lieferant: Dräger
X-am®
DRAE8323910EA 552 EUR
DRAE8323910
Flammable atmospheres detection device, Dräger X-am 2500 EX
Gas-Spürsysteme
The catalytic Ex sensor has high sensitivity to flammable gases and vapours, which is confirmed by the technical approval for measuring methane to nonane.
  • Audible and visual alarms
  • Gas entry from two sides
  • Large display for easy reading of values
The compact instrument was developed for personal protection.
Standards: IEC/EN 60079-29-1, Ex approval for zone 0.
Bestellinformation: For accessories and other models of X-am 2500, with additional or alternative sensors, please contact your local VWR agency.