(±)-3-Fluor-α-methylbenzylamin

Lieferant: SIA ENAMINE

ENAMEN30010651.1GEA 0 EUR
ENAMEN30010651.1G ENAMEN30010651.10G ENAMEN30010651.5G
(±)-3-Fluor-α-methylbenzylamin
(±)-3-Fluor-α-methylbenzylamin
Formel: C₈H₁₀FN
Molecular Weight: 139,17 g/mol
MDL: MFCD04038308
CAS-Nummer: 74788-45-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR