4-Isopropylbenzolsulfonylchlorid

Lieferant: SIA ENAMINE

Synonyme: 4-Isopropylbenzol-1-sulfonylchlorid4-Isopropylbenzolsulfonsäurechlorid4-Isopropylbenzolsulfochlorid

ENAMEN30005198.1GEA 0 EUR
ENAMEN30005198.1G ENAMEN30005198.5G ENAMEN30005198.10G
4-Isopropylbenzolsulfonylchlorid
4-Isopropylbenzolsulfonylchlorid
Formel: C₉H₁₁ClO₂S
Molecular Weight: 218,7 g/mol
Siedepunkt: 142…143 °C (1013 hPa)
Dichte: 1,225 g/cm³ (25 °C)
MDL: MFCD00041510
CAS-Nummer: 54997-90-9
UN: 3265
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR