4-Nitro-2-picolin-N-oxid

Lieferant: SIA ENAMINE

Synonyme: 2-Methyl-4-nitropyridin-1-oxid2-Methyl-4-nitropyridin-N-oxid4-Nitro-2-picolin-N-oxid

ENAMEN300105768.1GEA 0 EUR
ENAMEN300105768 ENAMEN300105768.5G ENAMEN300105768.1G
4-Nitro-2-picolin-N-oxid
4-Nitro-2-picolin-N-oxid
Formel: C₆H₆N₂O₃
Molecular Weight: 154,13 g/mol
Schmelzpunkt: 156 °C
MDL: MFCD00014625
CAS-Nummer: 5470-66-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR