4-(3,4-Dichlorphenoxy)piperidin

Lieferant: CHEMPUR

CHMP60-0139.1EA 0 EUR
CHMP60-0139.1 CHMP60-0139.500
4-(3,4-Dichlorphenoxy)piperidin
4-(3,4-Dichlorphenoxy)piperidin
Formel: C₁₁H₁₃Cl₂NO
MDL: MFCD04113542
CAS-Nummer: 245057-73-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR