2-(6-Bromo-2-naphthyl)-5-propyl-1,3,4-oxadiazol
Lieferant: COMBI-BLOCKS INC.
COBBHC-2562-1GEA
185
EUR
COBBHC-2562-1G
COBBHC-2562-5G
2-(6-Bromo-2-naphthyl)-5-propyl-1,3,4-oxadiazol
2-(6-Bromo-2-naphthyl)-5-propyl-1,3,4-oxadiazol
Formel:
C₁₅H₁₃BrN₂O |
MDL:
MFCD11855930 CAS-Nummer: 1133116-07-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...