1-Methyl-2-(tributylstannyl)pyrrol

Lieferant: CHEMPUR
Gefahr

CHMPFL11051.POREA 0 EUR
CHMPFL11051.POR
1-Methyl-2-(tributylstannyl)pyrrol
1-Methyl-2-(tributylstannyl)pyrrol
Formel: C₁₇H₃₃NSn
Molecular Weight: 370,15
MDL: MFCD01319022
CAS-Nummer: 118486-97-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR