1-Cycloheptenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS INC.

COBBPN-2525-5GEA 2260 EUR
COBBPN-2525-5G COBBPN-2525-1G
1-Cycloheptenylboronsäurepinakolester
1-Cycloheptenylboronsäurepinakolester
Formel: C₁₃H₂₃BO₂
MDL: MFCD10700152
CAS-Nummer: 287944-13-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR