4-(trans-4-Propylcyclohexyl)phenylboronsäure

Lieferant: COMBI-BLOCKS INC.

COBBBB-5913-1GEA 70.8 EUR
COBBBB-5913-1G
4-(trans-4-Propylcyclohexyl)phenylboronsäure
4-(trans-4-Propylcyclohexyl)phenylboronsäure
Formel: C₁₅H₂₃BO₂
Molecular Weight: 246,1529 g/mol
MDL: MFCD06658197
CAS-Nummer: 146862-02-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR